Assay Details
Gene Symbol: RBPMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655444.1 | XP_023511212.1 | 412 | 56 | ||
XM_023655452.1 | XP_023511220.1 | 414 | 56 | |||
GenBank mRNA | EF397515.1 | - | 3 - 4 | 168 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
