Assay Details
Gene Symbol: RPS27L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363667.4 | XP_003363715.1 | 264 | 83 | ||
GenBank mRNA | EF397516.1 | - | 1 - 2 | 103 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
