Assay Details
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145532.1 | NP_001139004.1 | 2 - 3 | 421 | 75 | |
XM_005596991.3 | XP_005597048.1 | 695 | 75 | |||
XM_005596992.3 | XP_005597049.1 | 722 | 75 | |||
XM_005596993.3 | XP_005597050.1 | 662 | 75 | |||
XM_005596994.2 | XP_005597051.1 | 643 | 75 | |||
XM_023651616.1 | XP_023507384.1 | 837 | 75 | |||
XM_023651617.1 | XP_023507385.1 | 532 | 75 | |||
GenBank mRNA | EF564279.2 | - | 2 - 3 | 421 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
