Assay Details
Gene Symbol: CYP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734095.2 | XP_014589581.2 | 1095 | 76 | ||
XM_023653657.1 | XP_023509425.1 | 1096 | 76 | |||
GenBank mRNA | EU220011.1 | - | 1 - 2 | 843 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
