Assay Details
Gene Symbol: ATP1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621035.1 | XP_023476803.1 | 699 | 106 | ||
GenBank mRNA | EU423858.1 | - | 4 - 5 | 516 | 106 | |
JL616041.1 | - | 4 - 5 | 535 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
