Assay Details
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163965.1 | NP_001157437.1 | 4 - 5 | 384 | 70 | |
XM_005601075.3 | XP_005601132.1 | 493 | 70 | |||
GenBank mRNA | M96552.1 | - | 4 - 5 | 326 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
