Assay Details
Gene Symbol: RGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144133.1 | NP_001137605.1 | 1 - 2 | 106 | 61 | |
GenBank mRNA | AB278562.1 | - | 1 - 2 | 106 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
