Assay Details
Gene Symbol: NREP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362841.4 | XP_003362889.1 | 453 | 92 | ||
XM_005599511.3 | XP_005599568.1 | 689 | 92 | |||
XM_023617869.1 | XP_023473637.1 | 269 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
