Assay Details
Gene Symbol: CNOT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488435.6 | XP_001488485.1 | 562 | 82 | ||
XM_023630446.1 | XP_023486214.1 | 586 | 82 | |||
GenBank mRNA | JL616284.2 | - | 3 - 4 | 488 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
