Assay Details
Gene Symbol: C13H16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616339.1 | XP_023472107.1 | 508 | 81 | ||
XM_023616340.1 | XP_023472108.1 | 511 | 81 | |||
GenBank mRNA | JL615893.1 | - | 1 - 2 | 190 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
