Assay Details
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242530.1 | NP_001229459.1 | 2 - 3 | 703 | 64 | |
XM_005608579.3 | XP_005608636.1 | 688 | 64 | |||
XM_014738513.2 | XP_014593999.1 | 495 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
