Assay Details
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495215.4 | XP_001495265.1 | 2038 | 68 | ||
XM_014730438.2 | XP_014585924.1 | 2073 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
