Assay Details
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601478.3 | XP_005601535.1 | 1939 | 68 | ||
XM_014732456.2 | XP_014587942.1 | 1765 | 68 | |||
XM_023622554.1 | XP_023478322.1 | 2162 | 68 | |||
XM_023622555.1 | XP_023478323.1 | 1937 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
