Assay Details
Gene Symbol: ADIPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163830.1 | NP_001157302.1 | 6 - 7 | 1110 | 79 | |
XM_005610811.3 | XP_005610868.1 | 1246 | 79 | |||
XM_014740477.2 | XP_014595963.1 | 873 | 79 | |||
XM_023642181.1 | XP_023497949.1 | 1263 | 79 | |||
GenBank mRNA | JL616067.2 | - | 4 - 5 | 727 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
