Assay Details
Gene Symbol: PTHLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163981.1 | NP_001157453.1 | 3 - 4 | 438 | 102 | |
XM_005610810.3 | XP_005610867.1 | 448 | 102 | |||
XM_014740461.2 | XP_014595947.1 | 586 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
