Assay Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205161.1 | NP_001192090.1 | 4 - 5 | 1247 | 69 | |
XM_014736158.2 | XP_014591644.1 | 1582 | 69 | |||
XM_014736161.2 | XP_014591647.1 | 1537 | 69 | |||
GenBank mRNA | GU982935.1 | - | 4 - 5 | 510 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
