Assay Details
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242477.1 | NP_001229406.1 | 4 - 5 | 426 | 90 | |
XM_023648289.1 | XP_023504057.1 | 562 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
