Assay Details
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242477.1 | NP_001229406.1 | 5 - 6 | 598 | 99 | |
XM_023648289.1 | XP_023504057.1 | 734 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
