Assay Details
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243145.1 | NP_001230074.1 | 2 - 3 | 769 | 60 | |
GenBank mRNA | CX601941.1 | - | 2 - 3 | 321 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
