Assay Details
Gene Symbol: SELENOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170338.1 | NP_001163809.1 | 3 - 4 | 591 | 94 | |
GenBank mRNA | CD464773.1 | - | 3 - 4 | 327 | 94 | |
CD536845.1 | - | 3 - 4 | 316 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
