Assay Details
Gene Symbol: SELENOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170422.2 | NP_001163893.1 | 4 - 5 | 220 | 59 | |
GenBank mRNA | CX605867.1 | - | 4 - 5 | 175 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
