Assay Details
Gene Symbol: SELENOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170422.2 | NP_001163893.1 | 5 - 6 | 316 | 108 | |
GenBank mRNA | CX605867.1 | - | 5 - 6 | 271 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
