Assay Details
Gene Symbol: SRSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195587.1 | NP_001182516.1 | 5 - 6 | 502 | 127 | |
XM_005605170.3 | XP_005605227.1 | 576 | 127 | |||
XM_005605171.3 | XP_005605228.1 | 519 | 127 | |||
XM_005605173.3 | XP_005605230.1 | 697 | 127 | |||
XM_014735590.2 | XP_014591076.1 | 801 | 127 | |||
XM_023627714.1 | XP_023483482.1 | 523 | 127 | |||
XM_023627715.1 | XP_023483483.1 | 699 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
