Assay Details
Gene Symbol: SRSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195588.1 | NP_001182517.1 | 3 - 4 | 514 | 62 | |
XM_005599771.3 | XP_005599828.1 | 499 | 62 | |||
XM_005599772.3 | XP_005599829.1 | 499 | 62 | |||
XM_005599773.3 | XP_005599830.1 | 499 | 62 | |||
GenBank mRNA | HQ889865.1 | - | 3 - 4 | 422 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
