Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197315.2 | NP_001184244.1 | 2 - 3 | 116 | 95 | |
XM_005600483.3 | XP_005600540.1 | 260 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
