Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197315.2 | NP_001184244.1 | 3 - 4 | 241 | 64 | |
XM_005600483.3 | XP_005600540.1 | 385 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
