Assay Details
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199127.1 | NP_001186056.1 | 3 - 4 | 301 | 63 | |
XM_005598730.3 | XP_005598787.1 | 578 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
