Assay Details
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204743.1 | NP_001191672.1 | 1 - 2 | 129 | 63 | |
XM_023651653.1 | XP_023507421.1 | 294 | 63 | |||
GenBank mRNA | DN511018.1 | - | 2 - 3 | 178 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
