Assay Details
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204844.2 | NP_001191773.1 | 2 - 3 | 2040 | 108 | |
XM_005599776.3 | XP_005599833.1 | 2136 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
