Assay Details
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204845.1 | NP_001191774.1 | 6 - 7 | 1007 | 67 | |
XM_005599777.3 | XP_005599834.1 | 972 | 67 | |||
XM_023618425.1 | XP_023474193.1 | 925 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
