Assay Details
Gene Symbol: CHTOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494814.5 | XP_001494864.1 | 733 | 75 | ||
XM_005610098.2 | XP_005610155.1 | 818 | 75 | |||
XM_005610100.2 | XP_005610157.1 | 730 | 75 | |||
XM_023641122.1 | XP_023496890.1 | 531 | 75 | |||
XM_023641123.1 | XP_023496891.1 | 531 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
