Assay Details
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606595.3 | XP_005606652.1 | 438 | 80 | ||
XM_014736813.2 | XP_014592299.1 | 465 | 80 | |||
XM_023631871.1 | XP_023487639.1 | 438 | 80 |
