Assay Details
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617146.1 | XP_023472914.1 | 333 | 78 | ||
XM_023617147.1 | XP_023472915.1 | 331 | 78 | |||
XM_023617148.1 | XP_023472916.1 | 332 | 78 | |||
XM_023617149.1 | XP_023472917.1 | 331 | 78 | |||
XM_023617150.1 | XP_023472918.1 | 333 | 78 | |||
XM_023617151.1 | XP_023472919.1 | 331 | 78 | |||
XM_023617152.1 | XP_023472920.1 | 329 | 78 | |||
XM_023617154.1 | XP_023472922.1 | 332 | 78 | |||
XM_023617155.1 | XP_023472923.1 | 287 | 78 | |||
XM_023617156.1 | XP_023472924.1 | 332 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
