Assay Details
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489408.6 | XP_001489458.1 | 220 | 114 | ||
XM_005601432.3 | XP_005601489.1 | 226 | 114 | |||
XM_023622707.1 | XP_023478475.1 | 223 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
