Assay Details
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504287.5 | XP_001504337.1 | 701 | 122 | ||
XM_005597648.3 | XP_005597705.1 | 660 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
