Assay Details
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309348.1 | NP_001296277.1 | 3 - 4 | 302 | 101 | |
XM_023644159.1 | XP_023499927.1 | 682 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
