Assay Details
Gene Symbol: RBBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634234.1 | XP_023490002.1 | 1099 | 66 | ||
XM_023634241.1 | XP_023490009.1 | 1238 | 66 | |||
XM_023634247.1 | XP_023490015.1 | 1099 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
