Assay Details
Gene Symbol: ATG4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490668.6 | XP_001490718.4 | 1111 | 81 | ||
XM_014729080.2 | XP_014584566.1 | 1040 | 81 | |||
XM_023633921.1 | XP_023489689.1 | 734 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
