Assay Details
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492446.5 | XP_001492496.1 | 346 | 78 | ||
XM_005602869.3 | XP_005602926.1 | 346 | 78 | |||
XM_005602870.3 | XP_005602927.1 | 349 | 78 | |||
XM_005602871.3 | XP_005602928.1 | 252 | 78 | |||
XM_014734045.2 | XP_014589531.1 | 386 | 78 | |||
XM_014734046.2 | XP_014589532.1 | 420 | 78 | |||
XM_014734047.2 | XP_014589533.1 | 414 | 78 | |||
XM_023652564.1 | XP_023508332.1 | 555 | 78 | |||
XM_023652590.1 | XP_023508358.1 | 315 | 78 | |||
XM_023652596.1 | XP_023508364.1 | 1556 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
