Assay Details
Gene Symbol: PAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504236.6 | XP_001504286.1 | 296 | 101 | ||
XM_014730642.2 | XP_014586128.1 | 348 | 101 | |||
XM_014730643.2 | XP_014586129.1 | 275 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
