Assay Details
Gene Symbol: MFRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503214.3 | XP_001503264.1 | 2634 | 62 | ||
XM_014741389.2 | XP_014596875.1 | 2636 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
