Assay Details
Gene Symbol: CD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253839.1 | NP_001240768.1 | 5 - 6 | 508 | 62 | |
GenBank mRNA | HQ890092.1 | - | 5 - 6 | 508 | 62 | |
JL616139.1 | - | 5 - 6 | 519 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
