Assay Details
Gene Symbol: KCNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256930.1 | NP_001243859.1 | 1 | 1723 | 75 | |
XM_005610813.2 | XP_005610870.1 | 1717 | 75 | |||
XM_023642184.1 | XP_023497952.1 | 1717 | 75 | |||
XM_023642185.1 | XP_023497953.1 | 1717 | 75 | |||
XM_023642186.1 | XP_023497954.1 | 1717 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
