Assay Details
Gene Symbol: NRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256955.1 | NP_001243884.1 | 1 | 3315 | 99 | |
XM_014736275.2 | XP_014591761.1 | 4162 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
