Assay Details
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267603.3 | NP_001254532.1 | 6 - 7 | 527 | 62 | |
XM_023626513.1 | XP_023482281.1 | 594 | 62 | |||
GenBank mRNA | JL616202.1 | - | 1 - 2 | 73 | 62 | |
JQ713943.1 | - | 6 - 7 | 530 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
