Assay Details
Gene Symbol: COX4I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270956.1 | NP_001257885.1 | 4 - 5 | 449 | 86 | |
XM_005604417.3 | XP_005604474.1 | 524 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
