Assay Details
Gene Symbol: CXCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278930.2 | NP_001265859.2 | 1 - 2 | 168 | 86 | |
GenBank mRNA | HQ889900.1 | - | 1 - 2 | 152 | 86 | |
HQ889980.1 | - | 1 - 2 | 106 | 86 | ||
HQ890098.1 | - | 1 - 2 | 76 | 86 | ||
JL616120.1 | - | 1 - 2 | 168 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
