Assay Details
Gene Symbol: TIPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283066.1 | NP_001269995.1 | 2 - 3 | 187 | 64 | |
XM_014733461.2 | XP_014588947.1 | 279 | 64 | |||
XM_014733462.2 | XP_014588948.1 | 344 | 64 | |||
XM_014733463.2 | XP_014588949.1 | 116 | 64 | |||
XM_023641395.1 | XP_023497163.1 | 616 | 64 | |||
XM_023641489.1 | XP_023497257.1 | 1596 | 64 | |||
GenBank mRNA | HQ890032.1 | - | 2 - 3 | 187 | 64 | |
JL638293.1 | - | 2 - 3 | 168 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014733463.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
