Assay Details
Gene Symbol: CALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283082.1 | NP_001270011.1 | 3 - 4 | 249 | 90 | |
GenBank mRNA | HQ889917.1 | - | 3 - 4 | 249 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
