Assay Details
Gene Symbol: LOC100146423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283088.1 | NP_001270017.1 | 18 - 19 | 2020 | 77 | |
XM_023621985.1 | XP_023477753.1 | 2476 | 77 | |||
XM_023621987.1 | XP_023477755.1 | 2476 | 77 | |||
XM_023621988.1 | XP_023477756.1 | 2475 | 77 | |||
XM_023621989.1 | XP_023477757.1 | 2478 | 77 | |||
XM_023621990.1 | XP_023477758.1 | 2469 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
